missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Phospho-APP/Protease Nexin II (T668) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF2508-SP
This item is not returnable.
View return policy
Description
APP Polyclonal specifically detects APP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| APP (p thr668) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APP | |
| Phosphopeptide containing the human APP/Protease Nexin II T668 site | |
| 25 μg | |
| Primary | |
| Detects multiple isoforms of human and mouse APP/Protease Nexin II when phosphorylated at sites corresponding to T668 of the human APP695 isoform. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 351 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction