missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FosB/G0S3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2214-SP
This item is not returnable.
View return policy
Description
FosB/G0S3 Monoclonal specifically detects FosB/G0S3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| FosB/G0S3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P53539 | |
| FOSB | |
| E. coli-derived recombinant human FosB Met1-Leu338 Accession # P53539 | |
| 25 μg | |
| Primary | |
| Detects human and mouse FosB in Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 269928 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activator protein 1, AP-1, DKFZp686C0818, FBJ murine osteosarcoma viral oncogene homolog B, G0S3, G0S3G0/G1 switch regulatory protein 3, GOS3, GOSB, MGC42291, oncogene FOS-B, protein fosB | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2354 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction