missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse DGK-epsilon Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7069-SP
This item is not returnable.
View return policy
Description
DGK-epsilon Polyclonal specifically detects DGK-epsilon in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DGK-epsilon | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DAG kinase epsilon, DAGK5, DAGK6, DGK, DGKE, DGKepsilon, DGK-epsilon, diacylglycerol kinase epsilon, diacylglycerol kinase, epsilon (64kD), diacylglycerol kinase, epsilon 64kDa, Diglyceride kinase epsilon, EC 2.7.1.107 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8526 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P52429 | |
| DGKE | |
| E. coli-derived recombinant human DGK-epsilon Asn314-Arg435 Accession # P52429 | |
| 25 μg | |
| Primary | |
| Detects recombinant human DGK-epsilon in direct ELISAs. Detects human and mouse DGK-epsilon in Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction