missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat RalA Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3270-SP
This item is not returnable.
View return policy
Description
RalA Monoclonal specifically detects RalA in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| RalA | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HINCONS, RAL, Rasl1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5898 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 399527 | |
| Western Blot 1 ug/mL | |
| P11233 | |
| RALA | |
| E. coli-derived recombinant human RalA Met1-Leu206 Accession # P11233 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat RalA in Western blots. In Western blots, no cross-reactivity with recombinant human RalB is observed. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction