missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Raf-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4540-SP
This item is not returnable.
View return policy
Description
Raf-1 Monoclonal specifically detects Raf-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Raf-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P04049 | |
| RAF1 | |
| E. coli-derived recombinant human Raf-1 Asn189-Thr353 Accession # P04049 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Raf-1 at 74 kDa in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 563002 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cRaf, c-Raf, c-Raf-1, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf1, Raf-1Oncogene RAF1, v-Raf-1, v-raf-1 murine leukemia viral oncogene homolog 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5894 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction