missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GABAB R1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marca: R&D Systems AF7000-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GABA-B R1 Polyclonal specifically detects GABA-B R1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| GABA-B R1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ92613, GABABR1, GABBR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, Gb1, GPRC3A, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2550 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9Z0U4 | |
| GABBR1 | |
| Chinese hamster ovary cell line CHO-derived recombinant rat GABA B R1 Gly17-Leu586 Accession # Q9Z0U4 | |
| 25 μg | |
| Primary | |
| Detects recombinant mouse and rat GABA B R1 in direct ELISAs and Western blots.Detects human and rat GABA B R1 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant rat GABA B R2 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido