missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken Netrin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF127-SP
This item is not returnable.
View return policy
Description
Netrin-2 Polyclonal specifically detects Netrin-2 in Chicken samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| Netrin-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Netrin2, NTN2L | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4917 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Q90923 | |
| NTN3 | |
| Mouse myeloma cell line NS0-derived recombinant chicken Netrin-2 Ala16-Pro581 Accession # Q90923 | |
| 25 μg | |
| Primary | |
| Detects chicken Netrin-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant chicken Netrin-1 is observed and less than 1% cross-reactivity with recombinant human Netrin-4, recombinant mouse (rm) Netrin-G1a and rmNetrin-G2a is observed. | |
| Chicken | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction