missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine SCF/c-kit Ligand Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2278-SP
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Canine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| SCF/c-kit Ligand | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4254 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.1-0.3 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q06220 | |
| KITLG | |
| E. coli-derived recombinant canine SCF/c-kit Ligand Lys26-Ala190 Accession # Q06220 | |
| 25 μg | |
| Primary | |
| Detects canine SCF/c-kit Ligand in ELISAs and Western blots. In sandwich immunoassays, less than 20% cross-reactivity with recombinant feline SCF is observed, less than 8% cross-reactivity with recombinant human SCF is observed, and less than 0.5% cross-reactivity with recombinant mouse SCF is observed. | |
| Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction