Learn More
Mouse CD4 Fluorescein-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems FAB554F-025
 View more versions of this product
Product Code. 18743558
  • £130.00 / 25 tests
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD4
Mouse CTL clone V4
25 Tests
Primary
Reacts with mouse CD4, an antigen co-receptor on the Tácell surface which interacts with MHCáII molecules on antigen presenting cells.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Flow Cytometry
GK1.5
Flow Cytometry 10 uL/10^6 cells
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
920
Mouse
Purified