missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human T-bet/TBX21 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC53851G-025
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 30009 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 525831 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| Q9UL17 | |
| TBX21 | |
| E. coli-derived recombinant human T-bet/TBX21 Glu326-Asn535 Accession # Q9UL17 | |
| 25 Tests | |
| Primary | |
| Detects human T-bet/TBX21 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction