missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for Glass Microanalysis Filter Holders

Accessories for filter holders for sample preparation.

Brand:  Merck XX1012502

287.54 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Additional Details : Weight : 0.11700kg

 View more versions of this product

Product Code. 15864146

  • £309.00 / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Accessories for filter holders for sample preparation.

Millipore Glass Vacuum Filtration Accessory ProductThese accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration. This item is an accessory part for the Millipore 25 mm Glass Microanalysis Filter Holder Kits.
Specifications

Specifications

Not Autoclavable
25 mm glass microanalysis vacuum filter
Non-sterile
25 mm
Base with stopper
Borosilicate Glass
Base with Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers