missing translation for 'onlineSavingsMsg'
Learn More

Windaus Ketone, TRC

Product Code. 30454408
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30454408

Brand: Toronto Research Chemicals TRCW49890025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Windaus Ketone
CAS 55812-80-1
Molecular Formula C19 H32 O
Quantity 25 mg
Synonym 4H-Inden-4-one','octahydro-7a-methyl-1-[(1R,2E,4R)-1,4,5-trimethyl-2-hexen-1-yl]-','(1R,3aR,7aR)-
InChI Formula InChI=1S/C19H32O/c1-13(2)14(3)8-9-15(4)16-10-11-17-18(20)7-6-12-19(16,17)5/h8-9,13-17H,6-7,10-12H2,1-5H3/b9-8+/t14-,15+,16+,17-,19+/m0/s1
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C(=O)CCC[C@]12C
IUPAC Name (1R,3aR,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-one
Molecular Weight (g/mol) 276.46
Formula Weight 276.2453
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.