missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valdecoxib, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Valdecoxib |
| CAS | 181695-72-7 |
| Molecular Formula | C16 H14 N2 O3 S |
| Quantity | 25 mg |
| Synonym | 4-(5-Methyl-3-phenyl-4-isoxazolyl)benzenesulfonamide, 4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonamide, Bextra, SC 65872, Valdecoxib, Valecoxib, Valus, Valz, Vx 2, Benzenesulfonamide','4-(5-methyl-3-phenyl-4-isoxazolyl)- |
| InChI Formula | InChI=1S/C16H14N2O3S/c1-11-15(12-7-9-14(10-8-12)22(17,19)20)16(18-21-11)13-5-3-2-4-6-13/h2-10H,1H3,(H2,17,19,20) |
| SMILES | Cc1onc(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)N |
| IUPAC Name | 4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 314.36 |
| Formula Weight | 314.0725 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction