missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylene-d12, TRC
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Specifications
Specifications
| Chemical Name or Material | Triphenylene-D12 |
| CAS | 17777-56-9 |
| Molecular Formula | C18 2H12 |
| Quantity | 25 mg |
| Synonym | Triphenylene-d12 (7CI,8CI,9CI), Triphenylene D12 |
| InChI Formula | InChI=1S/C18H12/c1-2-8-14-13(7-1)15-9-3-4-11-17(15)18-12-6-5-10-16(14)18/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D |
| SMILES | [2H]c1c([2H])c([2H])c2c3c([2H])c([2H])c([2H])c([2H])c3c4c([2H])c([2H])c([2H])c([2H])c4c2c1[2H] |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriotriphenylene |
| Molecular Weight (g/mol) | 240.36 |
| Formula Weight | 240.1692 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction