missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-trans-Verbenol, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | (-)-trans-Verbenol |
| CAS | 19890-02-9 |
| Molecular Formula | C10 H16 O |
| Quantity | 25 mg |
| Synonym | (1S,2R,5S)- 4,6,6-Trimethyl-bicyclo[3.1.1]hept-3-en-2-ol, (-)-(1S,4R,5S)-2-Pinen-4-ol, [1S-(1α,2α,5α)]-4,6,6-Trimethyl-bicyclo[3.1.1]hept-3-en-2-ol, (1S)-trans-Verbenol, (1S,4R,5S)-(-)-trans-Verbenol, (S)-(-)-trans-Verbenol, (S)-trans-Verbenol |
| InChI Formula | InChI=1S/C10H16O/c1-6-4-9(11)8-5-7(6)10(8,2)3/h4,7-9,11H,5H2,1-3H3/t7-,8+,9+/m0/s1 |
| SMILES | CC1=C[C@@H](O)[C@H]2C[C@@H]1C2(C)C |
| IUPAC Name | (1S,2R,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol |
| Molecular Weight (g/mol) | 152.233 |
| Formula Weight | 152.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction