missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene-d6 2,4-Di-iso-cyanate, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Toluene-d6 2,4-Di-iso-cyanate |
| CAS | 1398066-04-0 |
| Molecular Formula | C9 D6 N2 O2 |
| Quantity | 25 mg |
| Synonym | Toluene-d6 2,4-Di-iso-cyanate, 2,4-Diisocyanatotoluene-d6, 2,4-Diisocyanato-1-(methyl-d3)benzene-3,5,6-d3, 2,4-Toluenediisocyanate-D6 |
| InChI Formula | InChI=1S/C9H6N2O2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13/h2-4H,1H3/i1D3,2D,3D,4D |
| SMILES | [2H]c1c([2H])c(c(N=C=O)c([2H])c1N=C=O)C([2H])([2H])[2H] |
| IUPAC Name | 1,2,4-trideuterio-3,5-diisocyanato-6-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 180.1931 |
| Formula Weight | 180.0806 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction