missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Isotimolol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(S)-Isotimolol
Specifications
Specifications
| Chemical Name or Material | (S)-Isotimolol |
| CAS | 158636-96-5 |
| Analyte or Component Names | (S)-Isotimolol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H24 N4 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | (S)-Isotimolol, Timolol Maleate Impurity B as S-Enantiomer, Timolol Impurity B as S-Enantiomer, Timolol related compound B as S-Enantiomer, (2S)-3-[(1,1-dimethylethyl)amino]-2-[[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy]propan-1-ol |
| InChI Formula | InChI=1S/C13H24N4O3S/c1-13(2,3)14-8-10(9-18)20-12-11(15-21-16-12)17-4-6-19-7-5-17/h10,14,18H,4-9H2,1-3H3/t10-/m0/s1 |
| SMILES | CC(C)(C)NC[C@@H](CO)Oc1nsnc1N2CCOCC2 |
| IUPAC Name | (2S)-3-(tert-butylamino)-2-[(4-morpholin-4-yl-1,2,5-thiadiazol-3-yl)oxy]propan-1-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction