missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxybutynin EP Impurity D, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Oxybutynin EP Impurity D
Specifications
Specifications
| Chemical Name or Material | Phenylcyclohexylglycolic Acid |
| CAS | 4335-77-7 |
| Analyte or Component Names | Phenylcyclohexylglycolic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H18 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetic Acid (Phenylcyclohexylglycolic Acid), Oxybutynin Hydrochloride Imp. D (EP), Oxybutynin Imp. D (EP), Oxybutynin USP Related Compound A, Oxybutynin USP RC A, (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetic Acid, Phenylcyclohexylglycolic Acid, Oxybutynin Related Compound A, Oxybutynin Hydrochloride Impurity D, Oxybutynin Impurity D |
| InChI Formula | InChI=1S/C14H18O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12,17H,2,5-6,9-10H2,(H,15,16) |
| SMILES | OC(=O)C(O)(C1CCCCC1)c2ccccc2 |
| IUPAC Name | 2-cyclohexyl-2-hydroxy-2-phenylacetic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction