missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perphenazine Sulfone N1,N4-Dioxide Dihydrochloride (Perphenazine N1,N4,S,S-Tetraoxide Dihydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Perphenazine Sulfone N1,N4-Dioxide Dihydrochloride (Perphenazine N1,N4,S,S-Tetraoxide Dihydrochloride)
Specifications
Specifications
| Chemical Name or Material | Perphenazine Sulfone N1,N4-Dioxide Dihydrochloride |
| CAS | 2732347-57-6 |
| Analyte or Component Names | Perphenazine Sulfone N1,N4-Dioxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 Cl N3 O5 S . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Perphenazine Sulfone N1,N4-Dioxide Dihydrochloride, Perphenazine N1,N4,S,S-Tetraoxide Dihydrochloride |
| InChI Formula | InChI=1S/C21H26ClN3O5S.2ClH/c22-17-6-7-21-19(16-17)23(18-4-1-2-5-20(18)31(21,29)30)8-3-9-24(27)10-12-25(28,13-11-24)14-15-26;;/h1-2,4-7,16,26H,3,8-15H2;2*1H |
| SMILES | Cl.Cl.OCC[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3S(=O)(=O)c4ccc(Cl)cc24)CC1 |
| IUPAC Name | 2-[4-[3-(2-chloro-5,5-dioxophenothiazin-10-yl)propyl]-1,4-dioxidopiperazine-1,4-diium-1-yl]ethanol;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction