missing translation for 'onlineSavingsMsg'
Learn More

Oxolinic Acid Methyl Ester, Mikromol™

Product Code. 30051178
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30051178 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30051178

Brand: Mikromol MM0898.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxolinic Acid Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxolinic Acid Methyl Ester
CAS 50332-56-4
Analyte or Component Names Oxolinic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C14 H13 N O5
Certifications/Compliance ISO 9001
Synonym Oxolinic Acid Methyl Ester, 5-Ethyl-5,8-dihydro-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid methyl ester, Methyl 1-ethyl-6,7-methylenedioxy-4-quinolone-3-carboxylate
InChI Formula InChI=1S/C14H13NO5/c1-3-15-6-9(14(17)18-2)13(16)8-4-11-12(5-10(8)15)20-7-19-11/h4-6H,3,7H2,1-2H3
SMILES CCN1C=C(C(=O)OC)C(=O)c2cc3OCOc3cc12
IUPAC Name methyl 5-ethyl-8-oxo-[1,3]dioxolo[4,5-g]quinoline-7-carboxylate
Molecular Weight (g/mol) 275.26
Quantity 25 mg
Formula Weight 275.0794 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.