missing translation for 'onlineSavingsMsg'
Learn More

Oleoyl-L-carnitine, TRC

Product Code. 30008697
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008697 25 mg 25mg
30134377 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008697 Supplier Toronto Research Chemicals Supplier No. TRCO52670025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38677-66-6
Molecular Formula C25 H47 N O4
Molecular Weight (g/mol) 425.64
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z)-1-oxo-9-octadecen-1-yl]oxy]-, inner salt, (2R)-, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxo-9-octadecenyl)oxy]-, inner salt, [R-(Z)]-, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z)-1-oxo-9-octadecenyl]oxy]-, inner salt, (2R)- (9CI), C18:1 Carnitine, Oleoyl-L-carnitine, Oleoyl L-carnitine, Oleoylcarnitine
IUPAC Name (3R)-3-[(Z)-octadec-9-enoyl]oxy-4-(trimethylazaniumyl)butanoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H47NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4/h12-13,23H,5-11,14-22H2,1-4H3/b13-12-/t23-/m1/s1
Formula Weight 425.3505
Percent Purity >95%
Chemical Name or Material Oleoyl-L-carnitine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.