missing translation for 'onlineSavingsMsg'
Learn More

Acebutolol EP Impurity C (Acebutolol USP Related Compound A), Mikromol™

Product Code. 30050512
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30050512

Brand: Mikromol MM0435.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acebutolol EP Impurity C (Acebutolol USP Related Compound A)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Acetyl-4-hydroxyphenyl)butanamide
CAS 40188-45-2
Analyte or Component Names N-(3-Acetyl-4-hydroxyphenyl)butanamide
Shipping Condition Room Temperature
Molecular Formula C12 H15 N O3
Certifications/Compliance ISO 17025
Synonym Acebutolol Hydrochloride Imp. C (EP), Acebutolol Imp. C (EP), Acebutolol USP Related Compound A, Acebutolol USP RC A, N-(3-Acetyl-4-hydroxyphenyl)butanamide, Acebutolol Related Compound A, Acebutolol Hydrochloride Impurity C, Acebutolol Impurity C
InChI Formula InChI=1S/C12H15NO3/c1-3-4-12(16)13-9-5-6-11(15)10(7-9)8(2)14/h5-7,15H,3-4H2,1-2H3,(H,13,16)
SMILES CCCC(=O)Nc1ccc(O)c(c1)C(=O)C
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)butanamide
Molecular Weight (g/mol) 221.25
Quantity 25 mg
Formula Weight 221.1052 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.