missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acebutolol EP Impurity A, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Acebutolol EP Impurity A
Specifications
Specifications
| Chemical Name or Material | N-[3-Acetyl-4-[(2RS)-oxiran-2-ylmethoxy]phenyl]butanamide |
| CAS | 28197-66-2 |
| Analyte or Component Names | N-[3-Acetyl-4-[(2RS)-oxiran-2-ylmethoxy]phenyl]butanamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H19 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Acebutolol Hydrochloride Imp. A (EP), Acebutolol Imp. A (EP), N-[3-Acetyl-4-[(2RS)-oxiran-2-ylmethoxy]phenyl]butanamide, Acebutolol Hydrochloride Impurity A, Acebutolol Impurity A |
| InChI Formula | InChI=1S/C15H19NO4/c1-3-4-15(18)16-11-5-6-14(13(7-11)10(2)17)20-9-12-8-19-12/h5-7,12H,3-4,8-9H2,1-2H3,(H,16,18) |
| SMILES | CCCC(=O)Nc1ccc(OCC2CO2)c(c1)C(=O)C |
| IUPAC Name | N-[3-acetyl-4-(oxiran-2-ylmethoxy)phenyl]butanamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction