missing translation for 'onlineSavingsMsg'
Learn More

Melarsomine Dihydrochloride, TRC

Product Code. 30010879
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010879 25 mg 25mg
30012384 5 mg 5mg
30007970 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010879 Supplier Toronto Research Chemicals Supplier No. TRCM21510525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89141-50-4
Molecular Formula C13 H21 As N8 S2 . 2Cl H
Molecular Weight (g/mol) 501.33
Synonym Melarsenoxide Cysteamine, Immiticide, Cymelarsan, [4-[(4,6-Diamino-1,3,5-Triazin-2-yl)Amino]phenyl]-Bis(2-Aminoethyl) Ester Arsonodithious Acid Dihydrochloride
IUPAC Name N2-[4-[bis(2-aminoethylsulfanyl)arsanyl]phenyl]-1,3,5-triazine-2,4,6-triamine;dihydrochloride
SMILES Cl.Cl.NCCS[As](SCCN)c1ccc(Nc2nc(N)nc(N)n2)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C13H21AsN8S2.2ClH/c15-5-7-23-14(24-8-6-16)9-1-3-10(4-2-9)19-13-21-11(17)20-12(18)22-13;;/h1-4H,5-8,15-16H2,(H5,17,18,19,20,21,22);2*1H
Formula Weight 500.008
Chemical Name or Material Melarsomine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.