missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate (Ethyl Vincaminate), Mikromol™

Product Code. 30051507
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051507

Brand: Mikromol MM0406.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate (Ethyl Vincaminate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Vincaminate
CAS 40163-56-2
Analyte or Component Names Ethyl Vincaminate
Shipping Condition Room Temperature
Molecular Formula C22 H28 N2 O3
Certifications/Compliance ISO 17025
Synonym Vinpocetine Imp. A (EP), Vinpocetine USP Related Compound A, Vinpocetine USP RC A, Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]-naphthyridine-12-carboxylate, Ethyl Vincaminate, Vinpocetine Related Compound A, Vinpocetine Impurity A
InChI Formula InChI=1S/C22H28N2O3/c1-3-21-11-7-12-23-13-10-16-15-8-5-6-9-17(15)24(18(16)19(21)23)22(26,14-21)20(25)27-4-2/h5-6,8-9,19,26H,3-4,7,10-14H2,1-2H3/t19-,21+,22+/m1/s1
SMILES CCOC(=O)[C@@]1(O)C[C@]2(CC)CCCN3CCc4c([C@H]23)n1c5ccccc45
IUPAC Name Ethyl (12S,13aS,13bS)-13a-Ethyl-12-hydroxy-2,3,5,6,12,13,13a,13b-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]-naphthyridine-12-carboxylate
Molecular Weight (g/mol) 368.47
Quantity 25 mg
Formula Weight 368.21 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.