missing translation for 'onlineSavingsMsg'
Learn More

Etacrynic Acid Ethyl Ester, Mikromol™

Product Code. 30051812
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051812 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051812 Supplier Mikromol Supplier No. MM1207.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Etacrynic Acid Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etacrynic Acid Ethyl Ester
CAS 23870-41-9
Analyte or Component Names Etacrynic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C15 H16 Cl2 O4
Certifications/Compliance ISO 9001
Synonym Etacrynic Acid Ethyl Ester, 2-[2,3-dichloro-4-(2-methylene-1-oxobutyl)phenoxy]-Acetic acid ethyl ester, Acetic acid, [2,3-dichloro-4-(2-methylene-1-oxobutyl)phenoxy]-, ethyl ester (9CI), Acetic acid, [2,3-dichloro-4-(2-methylenebutyryl)phenoxy]-, ethyl ester (8CI), 2-[2,3-Dichloro-4-(2-methylenebutyryl)phenoxy]acetic acid ethyl ester
InChI Formula InChI=1S/C15H16Cl2O4/c1-4-9(3)15(19)10-6-7-11(14(17)13(10)16)21-8-12(18)20-5-2/h6-7H,3-5,8H2,1-2H3
SMILES CCOC(=O)COc1ccc(C(=O)C(=C)CC)c(Cl)c1Cl
IUPAC Name ethyl 2-[2,3-dichloro-4-(2-methylidenebutanoyl)phenoxy]acetate
Molecular Weight (g/mol) 331.19
Quantity 25 mg
Formula Weight 330.0426 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.