missing translation for 'onlineSavingsMsg'
Learn More

ent-Eletriptan, TRC

Product Code. 30009947
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009947 25 mg 25mg
30138331 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009947 Supplier Toronto Research Chemicals Supplier No. TRCE50500525MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1252673-29-2
Molecular Formula C22 H26 N2 O2 S
Molecular Weight (g/mol) 382.52
Synonym 3-[[(2S)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole;
IUPAC Name 5-[2-(benzenesulfonyl)ethyl]-3-[[(2S)-1-methylpyrrolidin-2-yl]methyl]-1H-indole
SMILES CN1CCC[C@H]1Cc2c[nH]c3ccc(CCS(=O)(=O)c4ccccc4)cc23

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H26N2O2S/c1-24-12-5-6-19(24)15-18-16-23-22-10-9-17(14-21(18)22)11-13-27(25,26)20-7-3-2-4-8-20/h2-4,7-10,14,16,19,23H,5-6,11-13,15H2,1H3/t19-/m0/s1
Formula Weight 382.1715
Chemical Name or Material ent-Eletriptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.