missing translation for 'onlineSavingsMsg'
Learn More

(E)-N-[3-(Methylamino)propyl]-3-(thiophen-2-yl)prop-2-enamide, Mikromol™

Product Code. 30051408
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30051408

Brand: Mikromol MM0449.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(E)-N-[3-(Methylamino)propyl]-3-(thiophen-2-yl)prop-2-enamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-N-[3-(Methylamino)propyl]-3-(thiophen-2-yl)prop-2-enamide
CAS 36700-39-7
Analyte or Component Names (E)-N-[3-(Methylamino)propyl]-3-(thiophen-2-yl)prop-2-enamide
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H16 N2 O S
Certifications/Compliance ISO 9001
Synonym Pyrantel Embonate Imp. B (EP), Pyrantel Embonate Impurity B
InChI Formula InChI=1S/C11H16N2OS/c1-12-7-3-8-13-11(14)6-5-10-4-2-9-15-10/h2,4-6,9,12H,3,7-8H2,1H3,(H,13,14)/b6-5+
SMILES CNCCCNC(=O)\C=C\c1cccs1
IUPAC Name (E)-N-[3-(methylamino)propyl]-3-thiophen-2-ylprop-2-enamide
Molecular Weight (g/mol) 224.32
Quantity 25 mg
Formula Weight 224.0983 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.