missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dosulepin Sulfoxide N-Oxide Hydrochloride (Dosulepin N,S-Dioxide Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Dosulepin Sulfoxide N-Oxide Hydrochloride (Dosulepin N,S-Dioxide Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Dosulepin Sulfoxide N-Oxide Hydrochloride |
| Analyte or Component Names | Dosulepin Sulfoxide N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H21 N O2 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dosulepin Sulfoxide N-Oxide Hydrochloride, Dosulepin N,S-Dioxide Hydrochloride |
| InChI Formula | InChI=1S/C19H21NO2S.ClH/c1-20(2,21)13-7-11-17-16-9-4-3-8-15(16)14-23(22)19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11+; |
| SMILES | Cl.C[N+](C)([O-])CC\C=C\1/c2ccccc2CS(=O)c3ccccc13 |
| IUPAC Name | (3E)-N,N-dimethyl-3-(5-oxo-6H-benzo[c][1]benzothiepin-11-ylidene)propan-1-amine oxide;hydrochloride |
| Molecular Weight (g/mol) | 363.9 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction