missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Despropylrotigotine ((6S)-5,6,7,8-Tetrahydro-6-[[2-(2-thienyl)ethyl]amino]-1-naphthalenol), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Despropylrotigotine ((6S)-5,6,7,8-Tetrahydro-6-[[2-(2-thienyl)ethyl]amino]-1-naphthalenol)
Specifications
Specifications
| Chemical Name or Material | Despropylrotigotine |
| CAS | 153409-14-4 |
| Analyte or Component Names | Despropylrotigotine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 N O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Despropylrotigotine, (6S)-5,6,7,8-Tetrahydro-6-[[2-(2-thienyl)ethyl]amino]-1-naphthalenol, (S)-5,6,7,8-Tetrahydro-6-[[2-(2-thienyl)ethyl]amino]-1-naphthalenol, Rotigotine EP impurity C, Rotigotine Impurity C |
| InChI Formula | InChI=1S/C16H19NOS/c18-16-5-1-3-12-11-13(6-7-15(12)16)17-9-8-14-4-2-10-19-14/h1-5,10,13,17-18H,6-9,11H2/t13-/m0/s1 |
| SMILES | Oc1cccc2C[C@H](CCc12)NCCc3cccs3 |
| IUPAC Name | (6S)-6-(2-thiophen-2-ylethylamino)-5,6,7,8-tetrahydronaphthalen-1-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction