missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Delta-Valerolactam-d4, TRC
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
Specifications
Specifications
| Chemical Name or Material | delta-Valerolactam-d4 |
| Molecular Formula | C5H5D4NO |
| Quantity | 25 mg |
| Synonym | 2-Piperidinone-d4, 2-Piperidone-d4, 2-Oxopiperidine-d4, 5-Pentanolactam-d4, 5-Amino-pentanoic Acid Lactam-d4, Valerolactim-d4, α-Piperidone-d4 |
| SMILES | O=C1CC([2H])([2H])C([2H])([2H])CN1 |
| Molecular Weight (g/mol) | 103.16 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction