missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Tocopherol-d3, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | beta-Tocopherol-d3 |
| CAS | 936230-75-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 H45 D3 O2 |
| Quantity | 25 mg |
| Synonym | (2R)-3,4-Dihydro-2,5,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol-d3, (+)- 2,5,8-Trimethyl-2-(4,8,12-trimethyltridecyl)-6-chromanol-d3, (R,R,R)-β-Tocopherol-d3, D-β-Tocopherol-d3, d-β-Tocopherol-d3, β-D-Tocopherol-d3 |
| InChI Formula | InChI=1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-24(6)26(29)19-23(5)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21-,22-,28-/m1/s1/i6D3 |
| SMILES | [2H]C([2H])([2H])c1c(O)cc(C)c2O[C@](C)(CCC[C@H](C)CCC[C@H](C)CCCC(C)C)CCc12 |
| IUPAC Name | (2R)-2,8-dimethyl-5-(trideuteriomethyl)-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol |
| Molecular Weight (g/mol) | 419.7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction