missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trospium EP Impurity A, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Trospium EP Impurity A
Specifications
Specifications
| Chemical Name or Material | Benzilic Acid |
| CAS | 76-93-7 |
| Analyte or Component Names | Benzilic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H12 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Trospium Chloride Imp. A (EP), Trospium Chloride USP Related Compound A, Trospium Chloride USP RC A, Hydroxydiphenylacetic Acid, Benzilic Acid, Trospium Chloride Imp. A (EP), Trospium Chloride Related Compound A, Trospium Related Compound A, Trospium Chloride Impurity A, Trospium Impurity A |
| InChI Formula | InChI=1S/C14H12O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,17H,(H,15,16) |
| SMILES | OC(=O)C(O)(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2-hydroxy-2,2-diphenylacetic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction