missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benoxaprofen 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Benoxaprofen 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers)
Specifications
Specifications
| Chemical Name or Material | Benoxaprofen 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers) |
| Analyte or Component Names | Benoxaprofen 1,3-Butylene Glycol Esters (Mixture of Regio- and Stereoisomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C20 H20 Cl N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benoxaprofen 1,3-Butylene Glycol Ester (Mixture of Isomers) |
| InChI Formula | InChI=1S/2C20H20ClNO4/c1-12(23)9-10-25-20(24)13(2)15-5-8-18-17(11-15)22-19(26-18)14-3-6-16(21)7-4-14;1-12(9-10-23)25-20(24)13(2)15-5-8-18-17(11-15)22-19(26-18)14-3-6-16(21)7-4-14/h2*3-8,11-13,23H,9-10H2,1-2H3 |
| SMILES | CC(O)CCOC(=O)C(C)c1ccc2oc(nc2c1)c3ccc(Cl)cc3.CC(CCO)OC(=O)C(C)c4ccc5oc(nc5c4)c6ccc(Cl)cc6 |
| IUPAC Name | 4-hydroxybutan-2-yl 2-[2-(4-chlorophenyl)-1,3-benzoxazol-5-yl]propanoate;3-hydroxybutyl 2-[2-(4-chlorophenyl)-1,3-benzoxazol-5-yl]propanoate |
| Molecular Weight (g/mol) | 747.66 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction