missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Descarboxamido-8-cyanotemozolomide (3,4-Dihydro-3-methyl-4-oxo-imidazo[5,1-d]-1,2,3,5-tetrazine-8-carbonitrile), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
8-Descarboxamido-8-cyanotemozolomide (3,4-Dihydro-3-methyl-4-oxo-imidazo[5,1-d]-1,2,3,5-tetrazine-8-carbonitrile)
Specifications
Specifications
| Chemical Name or Material | 8-Descarboxamido-8-cyanotemozolomide |
| CAS | 114601-31-9 |
| Analyte or Component Names | 8-Descarboxamido-8-cyanotemozolomide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H4 N6 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 8-Cyano-3-methyl-[3H]-imidazo[5,1-d]-tetrazin-4-one, Imidazo[5,1-d]-1,2,3,5-tetrazine-8-carbonitrile, 3,4-dihydro-3-methyl-4-oxo-, 8-Descarboxamido-8-cyano Temozolomide, Temozolomide Imp. C (EP), Temozolomide Impurity C |
| InChI Formula | InChI=1S/C6H4N6O/c1-11-6(13)12-3-8-4(2-7)5(12)9-10-11/h3H,1H3 |
| SMILES | CN1N=Nc2c(ncn2C1=O)C#N |
| IUPAC Name | 3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carbonitrile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction