missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2-phenoxyphenol, Mikromol™

Product Code. 30049668
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30049668

Brand: Mikromol MM0378.070025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Nitro-2-phenoxyphenol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitro-2-phenoxyphenol
CAS 70995-08-3
Analyte or Component Names 4-Nitro-2-phenoxyphenol
Shipping Condition Room Temperature
Molecular Formula C12 H9 N O4
Certifications/Compliance ISO 17025
Synonym 4-Nitro-2-phenoxyphenol, Nimesulide Imp. G (EP), Nimesulide Impurity G
InChI Formula InChI=1S/C12H9NO4/c14-11-7-6-9(13(15)16)8-12(11)17-10-4-2-1-3-5-10/h1-8,14H
SMILES Oc1ccc(cc1Oc2ccccc2)[N+](=O)[O-]
IUPAC Name 4-nitro-2-phenoxyphenol
Molecular Weight (g/mol) 231.2
Quantity 25 mg
Formula Weight 231.0532 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.