Learn More
Melford 4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulfate-β-D-glucopyranoside Sodium Salt
Product Code. 30018501
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 30018501
Brand: Melford M641500.025
Fluorescent substrate for N-Acetyl-β-D-glucosaminidase (Hexosaminidase A).
Fluorescent substrate for N-Acetyl-β-D-glucosaminidase (Hexosaminidase A). Allows more rapid detection of Tay-Sachs.
4-Methylumbelliferone < 50ppm
Nonsulfated Derivative: None Detected (NMR, TLC)
Checked by enzymatic assay
Specifications
Quantity 25 mg