missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonyl Chloride (Valdecoxib Sulfonyl Chloride), Mikromol™

Product Code. 30049995
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049995 25 mg 25mg
30049781 100 mg 100mg
30460644 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30049995 Supplier Mikromol Supplier No. MM0937.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonyl Chloride (Valdecoxib Sulfonyl Chloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonyl Chloride
CAS 509074-26-4
Analyte or Component Names 4-(5-Methyl-3-phenylisoxazol-4-yl)benzenesulfonyl Chloride
Shipping Condition Room Temperature
Molecular Formula C16 H12 Cl N O3 S
Certifications/Compliance ISO 9001
Synonym 4-(5-Methyl-3-phenyl-4-isoxazolyl)-benzenesulfonyl Chloride, 4-(5-Methyl-3-phenyl-4-isoxazolyl)benzenesulfonyl chloride
InChI Formula InChI=1S/C16H12ClNO3S/c1-11-15(12-7-9-14(10-8-12)22(17,19)20)16(18-21-11)13-5-3-2-4-6-13/h2-10H,1H3
SMILES Cc1onc(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)Cl
IUPAC Name 4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl chloride
Molecular Weight (g/mol) 333.79
Quantity 25 mg
Formula Weight 333.0226 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.