missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Epoxy-7-[(1S)-1-hydroxy-1,2,2-trimethylpropyl]-3,6-dimethoxy-6,14-ethano-14-morphinan-17-carbonitrile, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4,5-Epoxy-7-[(1S)-1-hydroxy-1,2,2-trimethylpropyl]-3,6-dimethoxy-6,14-ethano-14-morphinan-17-carbonitrile
Specifications
Specifications
| Chemical Name or Material | N-Cyano-3-O-methyl Norbuprenorphine |
| CAS | 16614-60-1 |
| Analyte or Component Names | N-Cyano-3-O-methyl Norbuprenorphine |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C27 H36 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (5α,7α)-4,5-Epoxy-18,19-dihydro-7-[(1S)-1-hydroxy-1,2,2-trimethylpropyl]-3,6-dimethoxy-6,14-ethenomorphinan-17-carbonitrile, 6,14-Ethenomorphinan-17-carbonitrile, 4,5-epoxy-18,19-dihydro-7-(1-hydroxy-1,2,2-trimethylpropyl)-3,6-dimethoxy-, [5α,7α(S)]- (9CI), 6,14-endo-Ethanotetrahydrothebaine, 17-cyano-17-demethyl-7α-(1-hydroxy-1,2,2-trimethylpropyl)- (8CI), N-Cyanonorbuprenorphine 3-methyl ether, Buprenorphine Imp. C (EP) |
| InChI Formula | InChI=1S/C27H36N2O4/c1-23(2,3)24(4,30)18-14-25-9-10-27(18,32-6)22-26(25)11-12-29(15-28)19(25)13-16-7-8-17(31-5)21(33-22)20(16)26/h7-8,18-19,22,30H,9-14H2,1-6H3/t18-,19-,22-,24+,25-,26+,27?/m1/s1 |
| SMILES | COc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)[C@]6(CC[C@@]35C[C@@H]6[C@](C)(O)C(C)(C)C)OC)C#N |
| Molecular Weight (g/mol) | 452.59 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction