missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Monohydrate (2-Azahypoxanthine Monohydrate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Monohydrate (2-Azahypoxanthine Monohydrate)
Specifications
Specifications
| Chemical Name or Material | 2-Azahypoxanthine Monohydrate |
| CAS | 3-3-7151 |
| Analyte or Component Names | 2-Azahypoxanthine Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H3 N5 O . H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dacarbazine Imp. A (EP), Temozolomide Imp. E (EP), Dacarbazine USP Related Compound B, Dacarbazine USP RC B, 3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Monohydrate, 2-Azahypoxanthine Monohydrate, Dacarbazine Related Compound B as Monohydrate, Dacarbazine Impurity A as Monohydrate, Temozolomide Impurity E as Monohydrate |
| InChI Formula | InChI=1S/C4H3N5O.H2O/c10-4-2-3(6-1-5-2)7-9-8-4;/h1H,(H2,5,6,7,8,10);1H2 |
| SMILES | O.O=C1NN=Nc2[nH]cnc12 |
| IUPAC Name | 3,7-dihydroimidazo[4,5-d]triazin-4-one;hydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction