missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoic Acid Ethyl Ester Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoic Acid Ethyl Ester Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2S)-2-[((1S)-1-(Ethoxycarbonyl)-3-phenylpropyl)amino]propanoic Acid Ethyl Ester Hydrochloride |
| CAS | 122375-72-8 |
| Analyte or Component Names | (2S)-2-[((1S)-1-(Ethoxycarbonyl)-3-phenylpropyl)amino]propanoic Acid Ethyl Ester Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H25 N O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]propanoic Acid Ethyl Ester Hydrochloride |
| InChI Formula | InChI=1S/C17H25NO4.ClH/c1-4-21-16(19)13(3)18-15(17(20)22-5-2)12-11-14-9-7-6-8-10-14;/h6-10,13,15,18H,4-5,11-12H2,1-3H3;1H/t13-,15-;/m0./s1 |
| SMILES | Cl.CCOC(=O)[C@H](C)N[C@@H](CCc1ccccc1)C(=O)OCC |
| IUPAC Name | ethyl (2S)-2-[[(2S)-1-ethoxy-1-oxopropan-2-yl]amino]-4-phenylbutanoate;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction