missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabeprazole EP Impurity H (Lansoprazole EP Impurity F ), Mikromol™
Description
Rabeprazole EP Impurity H (Lansoprazole EP Impurity F )
Specifications
Specifications
| Chemical Name or Material | 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole |
| CAS | 168167-42-8 |
| Analyte or Component Names | 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H12 Cl N3 O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Lansoprazole Imp. F (EP), Rabeprazole Sodium Imp. H (EP), Rabeprazole Imp. H (EP), Rabeprazole USP Related Compound F, Rabeprazole USP RC F, 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole, Rabeprazole Related Compound F, Lansoprazole Impurity F, Rabeprazole Sodium Impurity H, Rabeprazole Impurity H |
| InChI Formula | InChI=1S/C14H12ClN3OS/c1-9-10(15)6-7-16-13(9)8-20(19)14-17-11-4-2-3-5-12(11)18-14/h2-7H,8H2,1H3,(H,17,18) |
| SMILES | Cc1c(Cl)ccnc1CS(=O)c2nc3ccccc3[nH]2 |
| IUPAC Name | 2-[(4-chloro-3-methylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?