missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2-[(Diaminomethylidene)amino]thiazol-4-yl]methyl Carbamimidothioate Dihydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
[2-[(Diaminomethylidene)amino]thiazol-4-yl]methyl Carbamimidothioate Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | (2-[(Diaminomethylidene)amino]thiazol-4-yl)methyl Carbamimidothioate Dihydrochloride |
| CAS | 88046-01-9 |
| Analyte or Component Names | (2-[(Diaminomethylidene)amino]thiazol-4-yl)methyl Carbamimidothioate Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H10 N6 S2 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | [2-[(Diaminomethylidene)amino]thiazol-4-yl]methyl Carbamimidothioate Dihydrochloride, Famotidine Imp. H (EP), Carbamimidothioic acid, [2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl ester, dihydrochloride (9CI), [2-[(Diaminomethylidene)amino]thiazol-4-yl]methyl Carbamimidothioate Hydrochloride, S-(2-Guanidino-4-thiazolyl)methylisothiourea Dihydrochloride, Famotidine Imp. H (EP) as Hydrochloride, Famotidine Impurity H as Dihydrochloride |
| InChI Formula | InChI=1S/C6H10N6S2.2ClH/c7-4(8)12-6-11-3(2-14-6)1-13-5(9)10;;/h2H,1H2,(H3,9,10)(H4,7,8,11,12);2*1H |
| SMILES | Cl.Cl.NC(=N)Nc1nc(CSC(=N)N)cs1 |
| IUPAC Name | (2-carbamimidamido-1,3-thiazol-4-yl)methyl carbamimidothioate;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction