missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Amino-1-methyl-1H-benzimidazol-5-yl)phenylmethanone, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(2-Amino-1-methyl-1H-benzimidazol-5-yl)phenylmethanone
Specifications
Specifications
| Chemical Name or Material | (2-Amino-1-methyl-1H-benzimidazol-5-yl)phenylmethanone |
| CAS | 66066-76-0 |
| Analyte or Component Names | (2-Amino-1-methyl-1H-benzimidazol-5-yl)phenylmethanone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H13 N3 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Mebendazole Imp. C (EP), (2-Amino-1-methyl-1H-benzimidazol-5-yl)phenylmethanone, Mebendazole Related Compound C, Mebendazole Impurity C |
| InChI Formula | InChI=1S/C15H13N3O/c1-18-13-8-7-11(9-12(13)17-15(18)16)14(19)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17) |
| SMILES | Cn1c(N)nc2cc(ccc12)C(=O)c3ccccc3 |
| IUPAC Name | (2-amino-1-methylbenzimidazol-5-yl)-phenylmethanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction