missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,3S,5S)-2-Azabicyclo[3.1.0]hexane-3-carboxamide Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(1S,3S,5S)-2-Azabicyclo[3.1.0]hexane-3-carboxamide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (1S,3S,5S)-2-Azabicyclo[3.1.0]hexane-3-carboxamide Hydrochloride |
| CAS | 709031-39-0 |
| Analyte or Component Names | (1S,3S,5S)-2-Azabicyclo[3.1.0]hexane-3-carboxamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6H11ClN2O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Azabicyclo[3.1.0]hexane-3-carboxamide, hydrochloride (1:1), (1S,3S,5S)-, 2-Azabicyclo[3.1.0]hexane-3-carboxamide, monohydrochloride, (1S,3S,5S)- (9CI), (1S,3S,5S)-2-Azabicyclo[3.1.0]hexane-3-carboxamide hydrochloride |
| InChI Formula | InChI=1S/C6H10N2O.ClH/c7-6(9)5-2-3-1-4(3)8-5;/h3-5,8H,1-2H2,(H2,7,9);1H/t3-,4-,5-;/m0./s1 |
| SMILES | Cl.NC(=O)[C@@H]1C[C@@H]2C[C@@H]2N1 |
| IUPAC Name | (1S,3S,5S)-2-azabicyclo[3.1.0]hexane-3-carboxamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction