missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextropropoxyphene EP Impurity C (as Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
Conditionnement:
25mg
Description
Dextropropoxyphene EP Impurity C (as Hydrochloride)
Spécification
Spécification
| Chemical Name or Material | Butyroxyphene Hydrochloride |
| CAS | 2732346-98-2 |
| Analyte or Component Names | Butyroxyphene Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H31 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (1S,2R)-1-Benzyl-3-(dimethylamino)-2-methyl-1-phenylpropyl Butanoate Hydrochloride (Butyroxyphene Hydrochloride), Dextropropoxyphene Hydrochloride Imp. C (EP), Dextropropoxyphene Imp. C (EP), (1S,2R)-1-Benzyl-3-(dimethylamino)-2-methyl-1-phenylpropyl Butanoate Hydrochloride, Dextropropoxyphene Hydrochloride Imp. C (EP) as Hydrochloride, Butyroxyphene Hydrochloride, Dextropropoxyphene Hydrochloride Impurity C as Hydrochloride, Dextropropoxyphene Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C17H14N4O4/c1-25-17(24)21-16-19-13-7-4-11(8-14(13)20-16)15(23)10-2-5-12(6-3-10)18-9-22/h2-9H,1H3,(H,18,22)(H2,19,20,21,24) |
| SMILES | Cl.CCCC(=O)O[C@@](Cc1ccccc1)([C@H](C)CN(C)C)c2ccccc2 |
| IUPAC Name | [(2S,3R)-4-(dimethylamino)-3-methyl-1,2-diphenylbutan-2-yl] butanoate;hydrochloride |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu