missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1RS)-4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]-1-[4-(1-methylethyl)phenyl]butan-1-ol Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(1RS)-4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]-1-[4-(1-methylethyl)phenyl]butan-1-ol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 4-(Hydroxydiphenylmethyl)-alpha-[4-(1-methylethyl)phenyl]-1-piperidinebutanol hydrochloride |
| Analyte or Component Names | 4-(Hydroxydiphenylmethyl)-alpha-[4-(1-methylethyl)phenyl]-1-piperidinebutanol hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C31 H39 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-(Hydroxydiphenylmethyl)-α-[4-(1-methylethyl)phenyl]-1-piperidinebutanol hydrochloride, (1RS)-4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]-1-[4-(1-methylethyl)phenyl]butan-1-ol hydrochloride, ((+/-)-4-[1-hydroxy-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]-butyl]-isopropylbenzene, Fexofenadine related compound C, Fexofenadine Related Compound C as Hydrochloride, Fexofenadine Hydrochloride Impurity C as Hydrochloride, Fexofenadine Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C31H39NO2.ClH/c1-24(2)25-15-17-26(18-16-25)30(33)14-9-21-32-22-19-29(20-23-32)31(34,27-10-5-3-6-11-27)28-12-7-4-8-13-28;/h3-8,10-13,15-18,24,29-30,33-34H,9,14,19-23H2,1-2H3;1H |
| SMILES | Cl.CC(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4 |
| IUPAC Name | 4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]-1-(4-propan-2-ylphenyl)butan-1-ol;hydrochloride |
| Molecular Weight (g/mol) | 494.11 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction