missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17beta-Hydroxyandrosta-4,6-dien-3-one (Δ6-Testosterone), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
17beta-Hydroxyandrosta-4,6-dien-3-one (Δ6-Testosterone)
Specifications
Specifications
| Chemical Name or Material | Delta6-Testosterone |
| CAS | 2484-30-2 |
| Analyte or Component Names | Delta6-Testosterone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H26 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Testosterone Imp. I (EP), Testosterone USP Related Compound I, Testosterone USP RC I, 17beta-Hydroxyandrosta-4,6-dien-3-one, Delta6-Testosterone, Androstan-4,6-diene-17beta-ol-3-one, Testosterone Related Compound I, Testosterone Impurity I |
| SMILES | C[C@]12CC[C@H]3[C@@H](C=CC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O |
| IUPAC Name | (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimethyl-1,2,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 286.4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction