Learn More
Chem-Lab 25LT Hydrochloric acid 2 mol/l (72.922 g HCl / l
Brand: Chem-Lab CL05.0314.9025
25LT Hydrochloric acid 2 mol/l (72.922 g HCl / lH2O = 2 N (±0.004/20°C)), std sol
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrochloric acid 2 mol/l
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid