Learn More
Fisher Chemical 25KG AMMONIUMKLORID PSS 25KG
Brand: Fisher Chemical A/3880/70
25KG AMMONIUMKLORID PSS 25KG
Description
Safety and Handling
- Ammonium chloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.